3-(1,3-Dioxan-2-yl)propionic acid ≥97%

Supplier: Matrix Scientific

087015-1G 087015-2G 087015-5G
102851-986EA 735.38 USD
102851-986 102851-988 102851-990
3-(1,3-Dioxan-2-yl)propionic acid ≥97%
3-(1,3-Dioxan-2-yl)propionic acid

Hazard: Irritant

Formula: C₇H₁₂O₄
MDL Number: MFCD07700144
CAS Number: 774605-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR