5-Oxooctanoic acid ≥97%

Supplier: Matrix Scientific

087012-1G 087012-2G 087012-5G
102851-968EA 592.31 USD
102851-968 102851-970 102851-972
5-Oxooctanoic acid ≥97%
5-Oxooctanoic acid

Hazard: Irritant

Formula: C₈H₁₄O₃
MDL Number: MFCD01320168
CAS Number: 3637-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR