4-Oxoheptanoic acid ≥97%

Supplier: Matrix Scientific

087011-1G 087011-2G 087011-5G
102851-962EA 886.15 USD
102851-962 102851-964 102851-966
4-Oxoheptanoic acid ≥97%
4-Oxoheptanoic acid

Hazard: Irritant

Formula: C₇H₁₂O₃
MDL Number: MFCD01320167
CAS Number: 924-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR