5-Oxodecanoic acid ≥97%
Supplier: Matrix Scientific
087009-1G
087009-2G
087009-5G
102851-950EA
886.15
USD
102851-950
102851-952
102851-954
5-Oxodecanoic acid ≥97%
5-Oxodecanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₈O₃ |
MDL Number:
MFCD01320165 CAS Number: 624-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...