6-Oxodecanoic acid ≥97%

Supplier: Matrix Scientific

087007-1G 087007-2G 087007-5G
102851-938EA 590.77 USD
102851-938 102851-940 102851-942
6-Oxodecanoic acid ≥97%
6-Oxodecanoic acid

Hazard: Irritant

Formula: C₁₀H₁₈O₃
Melting Pt: 44…46 °C
MDL Number: MFCD01320163
CAS Number: 4144-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR