5-Oxononanoic acid ≥97%

Supplier: Matrix Scientific

087006-1G 087006-2G 087006-5G
102851-932EA 890.77 USD
102851-932 102851-934 102851-936
5-Oxononanoic acid ≥97%
5-Oxononanoic acid

Hazard: Irritant

Formula: C₉H₁₆O₃
MDL Number: MFCD01320162
CAS Number: 3637-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR