8-Oxodecanoic acid ≥97%

Supplier: Matrix Scientific

087005-1G 087005-2G 087005-5G
102851-926EA 592.31 USD
102851-926 102851-928 102851-930
8-Oxodecanoic acid ≥97%
8-Oxodecanoic acid

Hazard: Irritant

Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
MDL Number: MFCD01311672
CAS Number: 3006-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR