6-Oxooctanoic acid ≥97%

Supplier: Matrix Scientific

087003-1G 087003-2G 087003-5G
102851-914EA 592.31 USD
102851-914 102851-916 102851-918
6-Oxooctanoic acid ≥97%
6-Oxooctanoic acid

Hazard: Irritant

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD01320160
CAS Number: 4233-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR