5-Oxoheptanoic acid ≥97%
Supplier: Matrix Scientific
087002-1G
087002-2G
087002-5G
102851-908EA
958.46
USD
102851-908
102851-910
102851-912
5-Oxoheptanoic acid ≥97%
5-Oxoheptanoic acid
Hazard: Irritant
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol |
MDL Number:
MFCD01320159 CAS Number: 3637-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...