4-Oxohexanoic acid ≥97%

Supplier: Matrix Scientific

087001-1G 087001-2G 087001-5G
102851-902EA 886.15 USD
102851-902 102851-904 102851-906
4-Oxohexanoic acid ≥97%
4-Oxohexanoic acid

Hazard: Irritant

Formula: C₆H₁₀O₃
MDL Number: MFCD00068854
CAS Number: 1117-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR