6,6-Dimethyl-5-oxoheptanoic acid ≥97%

Supplier: Matrix Scientific

087000-1G 087000-2G 087000-5G
102851-896EA 592.31 USD
102851-896 102851-898 102851-900
6,6-Dimethyl-5-oxoheptanoic acid ≥97%
6,6-Dimethyl-5-oxoheptanoic acid

Hazard: Irritant

Formula: C₉H₁₆O₃
MW: 172.23 g/mol
MDL Number: MFCD01320156
CAS Number: 171557-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR