5,5-Dimethyl-4-oxohexanoic acid ≥97%

Supplier: Matrix Scientific

086999-1G 086999-2G 086999-5G
102851-890EA 886.15 USD
102851-890 102851-892 102851-894
5,5-Dimethyl-4-oxohexanoic acid ≥97%
5,5-Dimethyl-4-oxohexanoic acid

Hazard: Irritant

Formula: C₈H₁₄O₃
Melting Pt: 64…66 °C
MDL Number: MFCD01320155
CAS Number: 57965-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR