4-Cyclopropyl-4-oxobutyric acid ≥97%
Supplier: Matrix Scientific
086997-1G
086997-2G
086997-5G
102851-878EA
592.31
USD
102851-878
102851-880
102851-882
4-Cyclopropyl-4-oxobutyric acid ≥97%
4-Cyclopropyl-4-oxobutyric acid
Hazard: Irritant
Formula:
C₇H₁₀O₃ MW: 142.16 g/mol |
MDL Number:
MFCD01320151 CAS Number: 53712-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...