5-Cyclobutyl-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

086996-1G 086996-2G 086996-5G
102851-872EA 592.31 USD
102851-872 102851-874 102851-876
5-Cyclobutyl-5-oxovaleric acid ≥97%
5-Cyclobutyl-5-oxovaleric acid

Hazard: Irritant

Formula: C₉H₁₄O₃
MDL Number: MFCD01320148
CAS Number: 898766-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR