4-Cyclobutyl-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

086995-1G 086995-2G 086995-5G
102851-866EA 592.31 USD
102851-866 102851-868 102851-870
4-Cyclobutyl-4-oxobutyric acid ≥97%
4-Cyclobutyl-4-oxobutyric acid

Hazard: Irritant

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
MDL Number: MFCD01320147
CAS Number: 889953-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR