4-Cyclohexyl-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

086992-1G 086992-2G 086992-5G
102851-848EA 886.15 USD
102851-848 102851-850 102851-852
4-Cyclohexyl-4-oxobutyric acid ≥97%
4-Cyclohexyl-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₀H₁₆O₃
MDL Number: MFCD01320139
CAS Number: 15971-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR