2,2-Dimethyl-4-(4-methoxyphenyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

086990-1G 086990-2G 086990-5G
102851-836EA 795.38 USD
102851-836 102851-838 102851-840
2,2-Dimethyl-4-(4-methoxyphenyl)-4-oxobutyric acid ≥97%
2,2-Dimethyl-4-(4-methoxyphenyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₃H₁₆O₄
MDL Number: MFCD01320131
CAS Number: 15118-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR