6-(2-Fluorophenyl)-6-oxohexanoic acid ≥97%

Supplier: Matrix Scientific

086989-1G 086989-2G 086989-5G
102851-830EA 592.31 USD
102851-830 102851-832 102851-834
6-(2-Fluorophenyl)-6-oxohexanoic acid ≥97%
6-(2-Fluorophenyl)-6-oxohexanoic acid

Hazard: Irritant

Formula: C₁₂H₁₃FO₃
MDL Number: MFCD01320117
CAS Number: 56721-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR