5-(2-Fluorophenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-(2-Fluorophenyl)-5-oxopentanoic acid

086988-1G 086988-2G 086988-5G
102851-824EA 886.15 USD
102851-824 102851-826 102851-828
5-(2-Fluorophenyl)-5-oxovaleric acid ≥97%
5-(2-Fluorophenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₁H₁₁FO₃
MW: 210.21 g/mol
MDL Number: MFCD01320116
CAS Number: 199664-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR