4-(2-Naphthyl)-4-oxobutyric acid ≥97%

Supplier: Matrix Scientific

086985-1G 086985-2G 086985-5G
102851-806EA 490.77 USD
102851-806 102851-808 102851-810
4-(2-Naphthyl)-4-oxobutyric acid ≥97%
4-(2-Naphthyl)-4-oxobutyric acid

Hazard: Irritant

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD00090143
CAS Number: 1590-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR