8-(2-Methoxyphenyl)-8-oxooctanoic acid ≥97%

Supplier: Matrix Scientific

086984-1G 086984-2G 086984-5G
102851-800EA 592.31 USD
102851-800 102851-802 102851-804
8-(2-Methoxyphenyl)-8-oxooctanoic acid ≥97%
8-(2-Methoxyphenyl)-8-oxooctanoic acid

Hazard: Irritant

Formula: C₁₅H₂₀O₄
MDL Number: MFCD01311635
CAS Number: 898766-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR