6-(2-Methoxyphenyl)-6-oxohexanoic acid ≥97%

Supplier: Matrix Scientific

086983-1G 086983-2G 086983-5G
102851-794EA 592.31 USD
102851-794 102851-796 102851-798
6-(2-Methoxyphenyl)-6-oxohexanoic acid ≥97%
6-(2-Methoxyphenyl)-6-oxohexanoic acid

Hazard: Irritant

Formula: C₁₃H₁₆O₄
MDL Number: MFCD01311638
CAS Number: 107151-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR