4-(2-Methoxyphenyl)-4-oxobutyric acid ≥97%
Supplier: Matrix Scientific
086981-1G
086981-2G
086981-5G
102851-782EA
886.15
USD
102851-782
102851-784
102851-786
4-(2-Methoxyphenyl)-4-oxobutyric acid ≥97%
4-(2-Methoxyphenyl)-4-oxobutyric acid
Hazard: Irritant
Formula:
C₁₁H₁₂O₄ |
MDL Number:
MFCD01311637 CAS Number: 103987-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...