4-Oxo-4-(4-phenoxyphenyl)butyric acid ≥97%

Supplier: Matrix Scientific

086980-1G 086980-2G 086980-5G
102851-776EA 592.31 USD
102851-776 102851-778 102851-780
4-Oxo-4-(4-phenoxyphenyl)butyric acid ≥97%
4-Oxo-4-(4-phenoxyphenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₁₄O₄
MDL Number: MFCD00020539
CAS Number: 36330-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR