6-(3-Fluorophenyl)-6-oxohexanoic acid ≥97%

Supplier: Matrix Scientific

086979-1G 086979-2G 086979-5G
102851-770EA 592.31 USD
102851-770 102851-772 102851-774
6-(3-Fluorophenyl)-6-oxohexanoic acid ≥97%
6-(3-Fluorophenyl)-6-oxohexanoic acid

Hazard: Irritant

Formula: C₁₂H₁₃FO₃
MDL Number: MFCD01311618
CAS Number: 898765-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR