5-(3-Fluorophenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

086978-1G 086978-2G 086978-5G
102851-764EA 886.15 USD
102851-764 102851-766 102851-768
5-(3-Fluorophenyl)-5-oxovaleric acid ≥97%
5-(3-Fluorophenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₁H₁₁FO₃
MDL Number: MFCD01320063
CAS Number: 845790-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR