6-(3-Methoxyphenyl)-6-oxohexanoic acid ≥97%
Supplier: Matrix Scientific
086977-1G
086977-2G
086977-5G
102851-758EA
490.77
USD
102851-758
102851-760
102851-762
6-(3-Methoxyphenyl)-6-oxohexanoic acid ≥97%
6-(3-Methoxyphenyl)-6-oxohexanoic acid
Hazard: Irritant
Formula:
C₁₃H₁₆O₄ |
MDL Number:
MFCD01311623 CAS Number: 1884-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...