5-(3-Methylphenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

086975-1G 086975-2G 086975-5G
102851-746EA 490.77 USD
102851-746 102851-748 102851-750
5-(3-Methylphenyl)-5-oxovaleric acid ≥97%
5-(3-Methylphenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MDL Number: MFCD01320044
CAS Number: 36978-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR