2,2-Dimethyl-4-oxo-4-phenylbutyric acid ≥97%

Supplier: Matrix Scientific

086972-1G 086972-2G 086972-5G
102851-728EA 795.38 USD
102851-728 102851-730 102851-732
2,2-Dimethyl-4-oxo-4-phenylbutyric acid ≥97%
2,2-Dimethyl-4-oxo-4-phenylbutyric acid

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MDL Number: MFCD01320038
CAS Number: 15116-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR