6-(4-Methoxyphenyl)-6-oxohexanoic acid ≥97%

Supplier: Matrix Scientific

086971-1G 086971-2G 086971-5G
102851-722EA 490.77 USD
102851-722 102851-724 102851-726
6-(4-Methoxyphenyl)-6-oxohexanoic acid ≥97%
6-(4-Methoxyphenyl)-6-oxohexanoic acid

Hazard: Irritant

Formula: C₁₃H₁₆O₄
MDL Number: MFCD01311628
CAS Number: 5537-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR