7-(4-Chlorophenyl)-7-oxoheptanoic acid ≥97%
Supplier: Matrix Scientific
086969-1G
086969-2G
086969-5G
102851-710EA
490.77
USD
102851-710
102851-712
102851-714
7-(4-Chlorophenyl)-7-oxoheptanoic acid ≥97%
7-(4-Chlorophenyl)-7-oxoheptanoic acid
Hazard: Irritant
Formula:
C₁₃H₁₅ClO₃ MW: 254.72 g/mol |
MDL Number:
MFCD01320021 CAS Number: 35333-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...