7-(4-Fluorophenyl)-7-oxoheptanoic acid ≥97%

Supplier: Matrix Scientific

086968-1G 086968-2G 086968-5G
102851-704EA 490.77 USD
102851-704 102851-706 102851-708
7-(4-Fluorophenyl)-7-oxoheptanoic acid ≥97%
7-(4-Fluorophenyl)-7-oxoheptanoic acid

Hazard: Irritant

Formula: C₁₃H₁₅FO₃
MDL Number: MFCD01311598
CAS Number: 898787-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR