6-(4-Fluorophenyl)-6-oxohexanoic acid ≥97%
Supplier: Matrix Scientific
086967-1G
086967-2G
086967-5G
102851-698EA
490.77
USD
102851-698
102851-700
102851-702
6-(4-Fluorophenyl)-6-oxohexanoic acid ≥97%
6-(4-Fluorophenyl)-6-oxohexanoic acid
Hazard: Irritant
Formula:
C₁₂H₁₃FO₃ |
MDL Number:
MFCD01311599 CAS Number: 343319-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...