5-(3-Bromophenyl)-5-oxovaleric acid ≥97%

Supplier: Matrix Scientific

086965-1G 086965-5G
102851-686EA 735.38 USD
102851-686 102851-690
5-(3-Bromophenyl)-5-oxovaleric acid ≥97%
5-(3-Bromophenyl)-5-oxovaleric acid

Hazard: Irritant

Formula: C₁₁H₁₁BrO₃
MDL Number: MFCD00672058
CAS Number: 898787-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR