4'-Methoxy-2'-methyl-2,2,2-trifluoroacetophenone ≥97%
Supplier: Matrix Scientific
086962-1G
086962-2G
086962-5G
102851-668EA
347.69
USD
102851-668
102851-670
102851-672
4'-Methoxy-2'-methyl-2,2,2-trifluoroacetophenone ≥97%
4'-Methoxy-2'-methyl-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₁₀H₉F₃O₂ |
MDL Number:
MFCD03844198 CAS Number: 845823-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...