3-(4-Iodophenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086959-1G 086959-2G 086959-5G
102851-650EA 520 USD
102851-650 102851-652 102851-654
3-(4-Iodophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(4-Iodophenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₉H₆F₃IO
MDL Number: MFCD07700133
CAS Number: 898787-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR