3-(2-Iodophenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086958-1G 086958-2G 086958-5G
102851-644EA 541.54 USD
102851-644 102851-646 102851-648
3-(2-Iodophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(2-Iodophenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₉H₆F₃IO
MDL Number: MFCD07700131
CAS Number: 898787-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR