3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥95%

Supplier: Matrix Scientific

086957-1G 086957-2G 086957-5G
102851-638EA 520 USD
102851-638 102851-640 102851-642
3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥95%
3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant/Refrigerate

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD01764734
CAS Number: 22102-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR