3-(3-Methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086956-1G 086956-2G 086956-5G
102851-632EA 347.69 USD
102851-632 102851-634 102851-636
3-(3-Methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(3-Methoxyphenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD03844197
CAS Number: 22102-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR