3-(2-Methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086955-1G 086955-2G 086955-5G
102851-626EA 520 USD
102851-626 102851-628 102851-630
3-(2-Methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(2-Methoxyphenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant/Refrigerate

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD03844196
CAS Number: 870460-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR