3-(4-Methylphenyl)-1,1,1-trifluoro-2-propanone ≥97%
Supplier: Matrix Scientific
086954-1G
086954-2G
086954-5G
102851-620EA
520
USD
102851-620
102851-622
102851-624
3-(4-Methylphenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(4-Methylphenyl)-1,1,1-trifluoro-2-propanone
Hazard: Irritant
Formula:
C₁₀H₉F₃O |
MDL Number:
MFCD03844195 CAS Number: 75199-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...