3-(2-Methylphenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086952-1G 086952-2G 086952-5G
102851-608EA 347.69 USD
102851-608 102851-610 102851-612
3-(2-Methylphenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(2-Methylphenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₁₀H₉F₃O
MDL Number: MFCD03844193
CAS Number: 75199-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR