3-(4-Bromophenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086951-1G 086951-2G 086951-5G
102851-602EA 872.31 USD
102851-602 102851-604 102851-606
3-(4-Bromophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(4-Bromophenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₉H₆BrF₃O
MDL Number: MFCD03844192
CAS Number: 161809-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR