3-(3-Bromophenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(3-Bromophenyl)-1,1,1-trifluoropropan-2-one

086950-1G 086950-2G 086950-5G
102851-596EA 872.31 USD
102851-596 102851-598 102851-600
3-(3-Bromophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(3-Bromophenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₉H₆BrF₃O
MW: 267.05 g/mol
MDL Number: MFCD03844191
CAS Number: 898787-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR