3-(2-Bromophenyl)-1,1,1-trifluoro-2-propanone ≥97%
Supplier: Matrix Scientific
086949-1G
086949-2G
086949-5G
102851-590EA
872.31
USD
102851-590
102851-592
102851-594
3-(2-Bromophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(2-Bromophenyl)-1,1,1-trifluoro-2-propanone
Hazard: Irritant
Formula:
C₉H₆BrF₃O |
MDL Number:
MFCD03844190 CAS Number: 898787-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...