3-(4-Chlorophenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)-1,1,1-trifluoropropan-2-one

086948-1G 086948-2G 086948-5G
102851-584EA 872.31 USD
102851-584 102851-586 102851-588
3-(4-Chlorophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(4-Chlorophenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₉H₆ClF₃O
MW: 222.6 g/mol
MDL Number: MFCD03844189
CAS Number: 79611-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR