3-(2,5-Difluorophenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086942-1G 086942-2G 086942-5G
102851-554EA 469.23 USD
102851-554 102851-556 102851-558
3-(2,5-Difluorophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(2,5-Difluorophenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₉H₅F₅O
MDL Number: MFCD02260858
CAS Number: 898787-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR