3-(3,5-Difluorophenyl)-1,1,1-trifluoro-2-propanone ≥97%
Supplier: Matrix Scientific
086941-1G
086941-2G
086941-5G
102851-548EA
520
USD
102851-548
102851-550
102851-552
3-(3,5-Difluorophenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(3,5-Difluorophenyl)-1,1,1-trifluoro-2-propanone
Hazard: Irritant
Formula:
C₉H₅F₅O |
MDL Number:
MFCD02260857 CAS Number: 898787-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...