3-(5-Bromo-2-methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥97%

Supplier: Matrix Scientific

086939-1G 086939-2G 086939-5G
102851-536EA 552.31 USD
102851-536 102851-538 102851-540
3-(5-Bromo-2-methoxyphenyl)-1,1,1-trifluoro-2-propanone ≥97%
3-(5-Bromo-2-methoxyphenyl)-1,1,1-trifluoro-2-propanone

Hazard: Irritant

Formula: C₁₀H₈BrF₃O₂
MDL Number: MFCD07700129
CAS Number: 898787-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR