5'-Chloro-2'-methoxy-2,2,2-trifluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(5-Chloro-2-methoxyphenyl)-2,2,2-trifluoroethanone

086936-1G 086936-2G 086936-5G
102851-518EA 872.31 USD
102851-518 102851-520 102851-522
5'-Chloro-2'-methoxy-2,2,2-trifluoroacetophenone ≥97%
5'-Chloro-2'-methoxy-2,2,2-trifluoroacetophenone

Hazard: Irritant

Formula: C₉H₆ClF₃O₂
MW: 238.6 g/mol
Melting Pt: 101…104 °C
MDL Number: MFCD02260847
CAS Number: 886371-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR